Ethyl 2-(difluoromethyl)-8-fluoroimidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
PC301040-1G
PC301040-250MG
APOSPC301040-1GEA
522
EUR
APOSPC301040-1G
APOSPC301040-250MG
Ethyl 2-(difluoromethyl)-8-fluoroimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 2-(difluoromethyl)-8-fluoroimidazo[1,2-a]pyridine-3-carboxylate
Formula:
C₁₁H₉F₃N₂O₂ MW: 258.197 g/mol Melting Pt: 112…114 °C Storage Temperature: Ambient |
CAS Number:
2451256-49-6 |