2-(Difluoromethyl)-8-fluoroimidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
PC301041-1G PC301041-250MG
APOSPC301041-1GEA 522 EUR
APOSPC301041-1G APOSPC301041-250MG
2-(Difluoromethyl)-8-fluoroimidazo[1,2-a]pyridine-3-carboxylic acid
2-(Difluoromethyl)-8-fluoroimidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₉H₅F₃N₂O₂
MW: 230.143 g/mol
Melting Pt: 211…213 °C
Storage Temperature: Ambient
CAS Number: 2514942-09-5