8-Chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
PC301045-1G PC301045-250MG
APOSPC301045-1GEA 522 EUR
APOSPC301045-1G APOSPC301045-250MG
8-Chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
8-Chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₉H₅ClF₂N₂O₂
MW: 246.598 g/mol
Melting Pt: 211…213 °C
Storage Temperature: Ambient
CAS Number: 2514942-10-8