2-(Difluoromethyl)-8-methyl-2-imidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
PC301046-1G PC301046-250MG
APOSPC301046-1GEA 522 EUR
APOSPC301046-1G APOSPC301046-250MG
2-(Difluoromethyl)-8-methyl-2-imidazo[1,2-a]pyridine-3-carboxylic acid
2-(Difluoromethyl)-8-methyl-2-imidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₁₀H₈F₂N₂O₂
MW: 226.18 g/mol
Melting Pt: 201…203 °C
Storage Temperature: Ambient
CAS Number: 2514942-01-7