8-Bromo-6-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
PC301050-1G PC301050-250MG
APOSPC301050-1GEA 522 EUR
APOSPC301050-1G APOSPC301050-250MG
8-Bromo-6-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
8-Bromo-6-chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₉H₄BrClF₂N₂O₂
MW: 325.494 g/mol
Melting Pt: 205…207 °C
Storage Temperature: Ambient
CAS Number: 2514942-12-0