Ethyl 6,8-dichloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
PC301051-1G PC301051-250MG
APOSPC301051-1GEA 522 EUR
APOSPC301051-1G APOSPC301051-250MG
Ethyl 6,8-dichloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 6,8-dichloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₁H₈Cl₂F₂N₂O₂
MW: 309.096 g/mol
Melting Pt: 123…126 °C
Storage Temperature: Ambient
CAS Number: 2451256-45-2