6,8-Dichloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
PC301052-1G
PC301052-250MG
APOSPC301052-1GEA
522
EUR
APOSPC301052-1G
APOSPC301052-250MG
6,8-Dichloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
6,8-Dichloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Formula:
C₉H₄Cl₂F₂N₂O₂ MW: 281.043 g/mol Melting Pt: 213…215 °C Storage Temperature: Ambient |
CAS Number:
2451256-40-7 |