2-(Difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
PC301053-1G
PC301053-250MG
APOSPC301053-1GEA
522
EUR
APOSPC301053-1G
APOSPC301053-250MG
2-(Difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylic acid
2-(Difluoromethyl)-6-methylimidazo[1,2-a]pyridine-3-carboxylic acid
Formula:
C₁₀H₈F₂N₂O₂ MW: 226.18 g/mol Melting Pt: 211…213 °C Storage Temperature: Ambient |
CAS Number:
2168187-84-4 |