4-Bromo-2-fluoro-6-chlorobenzoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2-Chloro-6-fluorobenzoic acid
PC301202-1G PC301202-5G PC301202-10G
APOSPC301202-1GEA 27.7 EUR
APOSPC301202-1G APOSPC301202-5G APOSPC301202-10G
4-Bromo-2-fluoro-6-chlorobenzoic acid
4-Bromo-2-fluoro-6-chlorobenzoic acid
Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917163
CAS Number: 1321613-01-7