2,5-Dimethyl-1-[4-(trifluoromethyl)phenyl]-1H-pyrrole 97%
Supplier: Apollo Scientific
Warning
PC3017-1G PC3017-5G PC3017-25G
APOSPC3017-1GEA 55.1 EUR
APOSPC3017-1G APOSPC3017-5G APOSPC3017-25G
2,5-Dimethyl-1-[4-(trifluoromethyl)phenyl]-1H-pyrrole 97%
2,5-Dimethyl-1-[4-(trifluoromethyl)phenyl]-1H-pyrrole
Formula: C₁₃H₁₂F₃N
MW: 239.236 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00085042
CAS Number: 570-05-8