2-Chloro-4,5-difluorophenylacetonitrile 98%
Supplier: Apollo Scientific
PC302023-1G PC302023-5G PC302023-25G PC302023-100G
APOSPC302023-1GEA 105 EUR
APOSPC302023-1G APOSPC302023-5G APOSPC302023-25G APOSPC302023-100G
2-Chloro-4,5-difluorophenylacetonitrile 98%
2-Chloro-4,5-difluorophenylacetonitrile
Formula: C₈H₄ClF₂N
MW: 187.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660213
CAS Number: 874285-22-0
UN: 3439
ADR: 6.1,III