3-Chloro-2,4-difluorophenylacetonitrile 98%
Supplier: Apollo Scientific
Synonyms: 2-(3-Chloro-2,4-difluorophenyl)acetonitrile
PC302044-1G PC302044-5G PC302044-10G
APOSPC302044-1GEA 89.9 EUR
APOSPC302044-1G APOSPC302044-5G APOSPC302044-10G
3-Chloro-2,4-difluorophenylacetonitrile 98%
3-Chloro-2,4-difluorophenylacetonitrile
Formula: C₈H₄ClF₂N
MW: 187.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660338
CAS Number: 886761-67-7
UN: 3439
ADR: 6.1,III