5-Chloro-2-(trifluoromethyl)cinnamic acid 97%
Supplier: Apollo Scientific
PC302066-5G PC302066-1G
APOSPC302066-5GEA 224 EUR
APOSPC302066-5G APOSPC302066-1G
5-Chloro-2-(trifluoromethyl)cinnamic acid 97%
5-Chloro-2-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆ClF₃O₂
MW: 250.602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06205353
CAS Number: 773129-56-9