2,3-Difluoro-4-methyl-DL-phenylalanine
Supplier: Apollo Scientific
PC302121-1G
PC302121-5G
PC302121-10G
APOSPC302121-1GEA
163
EUR
APOSPC302121-1G
APOSPC302121-5G
APOSPC302121-10G
2,3-Difluoro-4-methyl-DL-phenylalanine
2,3-Difluoro-4-methyl-DL-phenylalanine
Formula:
C₁₀H₁₁F₂NO₂ MW: 215.197 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660244 CAS Number: 1215491-30-7 |