2-Chloro-3,6-difluoro-DL-phenylalanine
Supplier: Apollo Scientific
PC302125-1G PC302125-5G
APOSPC302125-1GEA 116 EUR
APOSPC302125-1G APOSPC302125-5G
2-Chloro-3,6-difluoro-DL-phenylalanine
2-Chloro-3,6-difluoro-DL-phenylalanine
Formula: C₉H₈ClF₂NO₂
MW: 235.615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116007
CAS Number: 1259981-12-8