2-Fluoro-5-(trifluoromethyl)-DL-phenylalanine
Supplier: Apollo Scientific
PC302134-1G PC302134-5G PC302134-10G
APOSPC302134-1GEA 116 EUR
APOSPC302134-1G APOSPC302134-5G APOSPC302134-10G
2-Fluoro-5-(trifluoromethyl)-DL-phenylalanine
2-Fluoro-5-(trifluoromethyl)-DL-phenylalanine
Formula: C₁₀H₉F₄NO₂
MW: 251.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115996
CAS Number: 1259994-87-0