4-Fluoro-3-(trifluoromethyl)-DL-phenylalanine
Supplier: Apollo Scientific
PC302149-1G PC302149-5G PC302149-10G
APOSPC302149-1GEA 128 EUR
APOSPC302149-1G APOSPC302149-5G APOSPC302149-10G
4-Fluoro-3-(trifluoromethyl)-DL-phenylalanine
4-Fluoro-3-(trifluoromethyl)-DL-phenylalanine
Formula: C₁₀H₉F₄NO₂
MW: 251.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116014
CAS Number: 1260004-58-7