2,6-Difluoro-3-methoxyphenylacetonitrile 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Cyanomethyl)-2,4-difluoroanisole
PC302162-1G PC302162-5G
APOSPC302162-1GEA 105 EUR
APOSPC302162-1G APOSPC302162-5G
2,6-Difluoro-3-methoxyphenylacetonitrile 98%
2,6-Difluoro-3-methoxyphenylacetonitrile
Formula: C₉H₇F₂NO
MW: 183.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115909
CAS Number: 886498-69-7
UN: 3439
ADR: 6.1 III