2-Methoxy-5-(trifluoromethoxy)benzoyl chloride
Supplier: Apollo Scientific
PC302177-10G PC302177-5G
APOSPC302177-10GEA 260 EUR
APOSPC302177-10G APOSPC302177-5G
2-Methoxy-5-(trifluoromethoxy)benzoyl chloride
2-Methoxy-5-(trifluoromethoxy)benzoyl chloride
Formula: C₉H₆ClF₃O₃
MW: 254.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115959
CAS Number: 191604-91-8