3-Fluoro-4-(trifluoromethoxy)cinnamic acid 98%
Supplier: Apollo Scientific
Warning
PC302200-1G PC302200-5G
APOSPC302200-1GEA 159 EUR
APOSPC302200-1G APOSPC302200-5G
3-Fluoro-4-(trifluoromethoxy)cinnamic acid 98%
3-Fluoro-4-(trifluoromethoxy)cinnamic acid
Formula: C₁₀H₆F₄O₃
MW: 250.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660209
CAS Number: 1261148-97-3