4,5-Difluoro-2-methoxycinnamic acid 97%
Supplier: Apollo Scientific
PC302204-1G PC302204-5G
APOSPC302204-1GEA 81.2 EUR
APOSPC302204-1G APOSPC302204-5G
4,5-Difluoro-2-methoxycinnamic acid 97%
4,5-Difluoro-2-methoxycinnamic acid
Formula: C₁₀H₈F₂O₃
MW: 214.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660221
CAS Number: 886499-69-0