4,5-Difluoro-2-methoxyphenylacetonitrile 98%
Supplier: Apollo Scientific
Synonyms: 2-(4,5-Difluoro-2-methoxyphenyl)acetonitrile
PC302205-1G PC302205-5G PC302205-10G
APOSPC302205-1GEA 109 EUR
APOSPC302205-1G APOSPC302205-5G APOSPC302205-10G
4,5-Difluoro-2-methoxyphenylacetonitrile 98%
4,5-Difluoro-2-methoxyphenylacetonitrile
Formula: C₉H₇F₂NO
MW: 183.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660349
CAS Number: 886761-74-6
UN: 2811
ADR: 6.1 III