6-Chloro-2-fluoro-3-methyl-DL-phenylalanine
Supplier: Apollo Scientific
PC302219-1G PC302219-5G PC302219-10G
APOSPC302219-1GEA 111 EUR
APOSPC302219-1G APOSPC302219-5G APOSPC302219-10G
6-Chloro-2-fluoro-3-methyl-DL-phenylalanine
6-Chloro-2-fluoro-3-methyl-DL-phenylalanine
Formula: C₁₀H₁₁ClFNO₂
MW: 231.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660188
CAS Number: 1259982-09-6