2-Bromo-3,6-difluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC302225-1G PC302225-5G
APOSPC302225-1GEA 75.4 EUR
APOSPC302225-1G APOSPC302225-5G
2-Bromo-3,6-difluorobenzoic acid 97%
2-Bromo-3,6-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11849930
CAS Number: 124244-65-1