2-Methoxy-5-(trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
PC302258-1G PC302258-5G PC302258-10G
APOSPC302258-1GEA 122 EUR
APOSPC302258-1G APOSPC302258-5G APOSPC302258-10G
2-Methoxy-5-(trifluoromethyl)phenylacetonitrile 97%
2-Methoxy-5-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₈F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09832243
CAS Number: 1000542-90-4
UN: 3439
ADR: 6.1,III