3-(2,3-Difluoro-4-methylphenyl)propionic acid 98%
Supplier: Apollo Scientific
PC302263-1G PC302263-5G
APOSPC302263-1GEA 214 EUR
APOSPC302263-1G APOSPC302263-5G
3-(2,3-Difluoro-4-methylphenyl)propionic acid 98%
3-(2,3-Difluoro-4-methylphenyl)propionic acid
Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832372
CAS Number: 1017779-58-6