3-Ethoxy-2,4-difluorophenylacetonitrile 97%
Supplier: Apollo Scientific
PC302294-1G PC302294-5G PC302294-10G
APOSPC302294-1GEA 116 EUR
APOSPC302294-1G APOSPC302294-5G APOSPC302294-10G
3-Ethoxy-2,4-difluorophenylacetonitrile 97%
3-Ethoxy-2,4-difluorophenylacetonitrile
Formula: C₁₀H₉F₂NO
MW: 197.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258730
CAS Number: 1017778-26-5
UN: 3276
ADR: 6.1,III