3-Fluoro-2-methoxycinnamic acid 98%
Supplier: Apollo Scientific
PC302299-1G PC302299-5G
APOSPC302299-1GEA 87 EUR
APOSPC302299-1G APOSPC302299-5G
3-Fluoro-2-methoxycinnamic acid 98%
3-Fluoro-2-methoxycinnamic acid
Formula: C₁₀H₉FO₃
MW: 196.175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999421
CAS Number: 1092460-67-7