4-Chloro-3,5-difluorocinnamic acid 97%
Supplier: Apollo Scientific
PC302325-1G PC302325-5G
APOSPC302325-1GEA 270 EUR
APOSPC302325-1G APOSPC302325-5G
4-Chloro-3,5-difluorocinnamic acid 97%
4-Chloro-3,5-difluorocinnamic acid
Formula: C₉H₅ClF₂O₂
MW: 218.585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23795326
CAS Number: 1163258-83-0