6-Bromo-2,3-difluorobenzoic acid
Supplier: Apollo Scientific
PC302365-1G PC302365-5G
APOSPC302365-1GEA 102 EUR
APOSPC302365-1G APOSPC302365-5G
6-Bromo-2,3-difluorobenzoic acid
6-Bromo-2,3-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
MW: 236.998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368092
CAS Number: 183065-72-7