2,3-Difluoro-4-methoxybenzoyl chloride
Supplier: Apollo Scientific
PC302372-10G PC302372-5G
APOSPC302372-10GEA 440 EUR
APOSPC302372-10G APOSPC302372-5G
2,3-Difluoro-4-methoxybenzoyl chloride
2,3-Difluoro-4-methoxybenzoyl chloride
Formula: C₈H₅ClF₂O₂
MW: 206.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115982
CAS Number: 849632-69-5