2-Chloro-3-(trifluoromethyl)benzamide 97%
Supplier: Apollo Scientific
PC302383-5G PC302383-1G PC302383-25G
APOSPC302383-5GEA 105 EUR
APOSPC302383-5G APOSPC302383-1G APOSPC302383-25G
2-Chloro-3-(trifluoromethyl)benzamide 97%
2-Chloro-3-(trifluoromethyl)benzamide
Formula: C₈H₅ClF₃NO
MW: 223.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115831
CAS Number: 39959-94-9