2-Methoxy-5-(trifluoromethyl)benzamide 97%
Supplier: Apollo Scientific
Warning
PC302390-5G PC302390-1G
APOSPC302390-5GEA 148 EUR
APOSPC302390-5G APOSPC302390-1G
2-Methoxy-5-(trifluoromethyl)benzamide 97%
2-Methoxy-5-(trifluoromethyl)benzamide
Formula: C₉H₈F₃NO₂
MW: 219.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06409083
CAS Number: 874804-06-5
UN: 2811
ADR: 6.1 III