2,4-Difluoro-3-methoxy-DL-phenylalanine
Supplier: Apollo Scientific
PC302439-1G PC302439-5G
APOSPC302439-1GEA 116 EUR
APOSPC302439-1G APOSPC302439-5G
2,4-Difluoro-3-methoxy-DL-phenylalanine
2,4-Difluoro-3-methoxy-DL-phenylalanine
Formula: C₁₀H₁₁F₂NO₃
MW: 231.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832238
CAS Number: 1259975-67-1