2,6-Difluoro-3-methoxy-DL-phenylalanine
Supplier: Apollo Scientific
PC302441-1G PC302441-5G
APOSPC302441-1GEA 136 EUR
APOSPC302441-1G APOSPC302441-5G
2,6-Difluoro-3-methoxy-DL-phenylalanine
2,6-Difluoro-3-methoxy-DL-phenylalanine
Formula: C₁₀H₁₁F₂NO₃
MW: 231.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832371
CAS Number: 1256482-66-2