2-Methoxy-4-(trifluoromethoxy)benzamide
Supplier: Apollo Scientific
PC302463-10G PC302463-5G
APOSPC302463-10GEA 395 EUR
APOSPC302463-10G APOSPC302463-5G
2-Methoxy-4-(trifluoromethoxy)benzamide
2-Methoxy-4-(trifluoromethoxy)benzamide
Formula: C₉H₈F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD06660245
CAS Number: 886500-76-1