2-Methoxy-5-(trifluoromethyl)benzyl alcohol 98%
Supplier: Apollo Scientific
Warning
PC302468-10G PC302468-5G
APOSPC302468-10GEA 191 EUR
APOSPC302468-10G APOSPC302468-5G
2-Methoxy-5-(trifluoromethyl)benzyl alcohol 98%
2-Methoxy-5-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O₂
MW: 206.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203094
CAS Number: 685126-89-0