3-[3-Chloro-5-(trifluoromethyl)phenyl]propionic acid 98%
Supplier: Apollo Scientific
Synonyms: 3-(3-Chloro-5-(trifluoromethyl)phenyl)propanoic acid
PC302507-1G PC302507-5G PC302507-10G PC302507-25G
APOSPC302507-1GEA 125 EUR
APOSPC302507-1G APOSPC302507-5G APOSPC302507-10G APOSPC302507-25G
3-[3-Chloro-5-(trifluoromethyl)phenyl]propionic acid 98%
3-[3-Chloro-5-(trifluoromethyl)phenyl]propionic acid
Formula: C₁₀H₈ClF₃O₂
MW: 252.618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025401
CAS Number: 916420-78-5