3-Ethoxy-2,6-difluorobenzamide 98%
Supplier: Apollo Scientific
PC302546-10G PC302546-5G PC302546-1G
APOSPC302546-10GEA 224 EUR
APOSPC302546-10G APOSPC302546-5G APOSPC302546-1G
3-Ethoxy-2,6-difluorobenzamide 98%
3-Ethoxy-2,6-difluorobenzamide
Formula: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD11519367
CAS Number: 1092461-26-1