3-Methoxy-2-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
PC302557-1G PC302557-5G
APOSPC302557-1GEA 159 EUR
APOSPC302557-1G APOSPC302557-5G
3-Methoxy-2-(trifluoromethyl)benzoyl chloride 97%
3-Methoxy-2-(trifluoromethyl)benzoyl chloride
Formula: C₉H₆ClF₃O₂
MW: 238.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398586
CAS Number: 1261618-61-4