4-Fluoro-3-(trifluoromethoxy)benzamide 98%
Supplier: Apollo Scientific
PC302594-1G PC302594-5G
APOSPC302594-1GEA 180 EUR
APOSPC302594-1G APOSPC302594-5G
4-Fluoro-3-(trifluoromethoxy)benzamide 98%
4-Fluoro-3-(trifluoromethoxy)benzamide
Formula: C₈H₅F₄NO₂
MW: 223.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07772784
CAS Number: 1206593-25-0