5-Fluoro-2-(trifluoromethyl)-DL-phenylalanine
Supplier: Apollo Scientific
PC302620-1G PC302620-5G PC302620-10G
APOSPC302620-1GEA 116 EUR
APOSPC302620-1G APOSPC302620-5G APOSPC302620-10G
5-Fluoro-2-(trifluoromethyl)-DL-phenylalanine
5-Fluoro-2-(trifluoromethyl)-DL-phenylalanine
Formula: C₁₀H₉F₄NO₂
MW: 251.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116013
CAS Number: 1260002-41-2