2,4-Difluoro-3-methoxybenzoyl chloride 98%
Supplier: Apollo Scientific
PC302662-25G PC302662-5G PC302662-1G
APOSPC302662-25GEA 1350 EUR
APOSPC302662-25G APOSPC302662-5G APOSPC302662-1G
2,4-Difluoro-3-methoxybenzoyl chloride 98%
2,4-Difluoro-3-methoxybenzoyl chloride
Formula: C₈H₅ClF₂O₂
MW: 206.574 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115933
CAS Number: 221221-11-0