2-Ethoxy-3,5-difluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC302678-10G PC302678-5G PC302678-1G
APOSPC302678-10GEA 282 EUR
APOSPC302678-10G APOSPC302678-5G APOSPC302678-1G
2-Ethoxy-3,5-difluorobenzoic acid 98%
2-Ethoxy-3,5-difluorobenzoic acid
Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258711
CAS Number: 1017779-70-2