2-Methoxy-5-(trifluoromethyl)phenylacetic acid 97%
Supplier: Apollo Scientific
Warning
PC302695-1G PC302695-5G
APOSPC302695-1GEA 180 EUR
APOSPC302695-1G APOSPC302695-5G
2-Methoxy-5-(trifluoromethyl)phenylacetic acid 97%
2-Methoxy-5-(trifluoromethyl)phenylacetic acid
Formula: C₁₀H₉F₃O₃
MW: 234.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832244
CAS Number: 1000523-82-9