3-Chloro-5-fluoro-4-methoxybenzoyl chloride
Supplier: Apollo Scientific
Danger
PC302717-10G PC302717-5G
APOSPC302717-10GEA 328 EUR
APOSPC302717-10G APOSPC302717-5G
3-Chloro-5-fluoro-4-methoxybenzoyl chloride
3-Chloro-5-fluoro-4-methoxybenzoyl chloride
Formula: C₈H₅Cl₂FO₂
MW: 223.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115865
CAS Number: 886497-26-3
UN: 3261
ADR: 8 III