3-Fluoro-5-methylbenzoyl chloride
Supplier: Apollo Scientific
PC302727-10G PC302727-5G PC302727-1G
APOSPC302727-10GEA 250 EUR
APOSPC302727-10G APOSPC302727-5G APOSPC302727-1G
3-Fluoro-5-methylbenzoyl chloride
3-Fluoro-5-methylbenzoyl chloride
Formula: C₈H₆ClFO
MW: 172.584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115884
CAS Number: 886497-77-4
UN: 3265
ADR: 8,III