4,5-Difluoro-2-methoxybenzamide 97%
Supplier: Apollo Scientific
PC302734-5G PC302734-1G
APOSPC302734-5GEA 136 EUR
APOSPC302734-5G APOSPC302734-1G
4,5-Difluoro-2-methoxybenzamide 97%
4,5-Difluoro-2-methoxybenzamide
Formula: C₈H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06660345
CAS Number: 425702-23-4