2-Fluoro-5-methylbenzyl alcohol
Supplier: Apollo Scientific
Warning
PC302767-10G PC302767-5G
APOSPC302767-10GEA 351 EUR
APOSPC302767-10G APOSPC302767-5G
2-Fluoro-5-methylbenzyl alcohol
2-Fluoro-5-methylbenzyl alcohol
Formula: C₈H₉FO
MW: 140.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06202823
CAS Number: 64977-30-6