3-Fluoro-4-(trifluoromethoxy)phenylacetonitrile 98%
Supplier: Apollo Scientific
Warning
PC302843-1G PC302843-5G PC302843-10G
APOSPC302843-1GEA 163 EUR
APOSPC302843-1G APOSPC302843-5G APOSPC302843-10G
3-Fluoro-4-(trifluoromethoxy)phenylacetonitrile 98%
3-Fluoro-4-(trifluoromethoxy)phenylacetonitrile
Formula: C₉H₅F₄NO
MW: 219.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660344
CAS Number: 886761-71-3